Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igbp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015777.2 | NP_056592.2 | 1 | 575 | ||
GenBank mRNA | AJ010637.1 | - | 1 | 575 | ||
BC125601.1 | - | 1 | 675 | 40130116 | ||
BC131968.1 | - | 1 | 675 | 40130245 |