Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh2b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK140319.1 | - | 9 | 2232 | ||
AK140325.1 | - | 9 | 2232 | |||
BC057334.1 | - | 9 | 2284 | 6811850 |