Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021429.3 | NP_067404.2 | 7 | 1752 | ||
GenBank mRNA | AJ132192.1 | - | 8 | 1245 | ||
AK040654.1 | - | 7 | 1753 | |||
AK084802.1 | - | 7 | 1744 | |||
AK154696.1 | - | 7 | 1748 | |||
AK170838.1 | - | 7 | 1739 | |||
BC095977.1 | - | 7 | 1749 |