Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cd209e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_130905.2 | NP_570975.1 | 6 | 1087 | ||
GenBank mRNA | AF373412.1 | - | 6 | 1087 | ||
AK040917.1 | - | 6 | 1109 |