Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam83f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145986.2 | NP_666098.2 | 5 | 2296 | ||
GenBank mRNA | AK030342.1 | - | 5 | 2278 | ||
AK145865.1 | - | 5 | 2296 | |||
BC016600.1 | - | 5 | 2274 | 3153977 |