Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amigo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004293.2 | NP_001004293.1 | 2 | 956 | ||
NM_001287093.1 | NP_001274022.1 | 2 | 956 | |||
NM_146137.3 | NP_666249.2 | 2 | 991 | |||
GenBank mRNA | AB167509.1 | - | 1 | 612 | ||
AK035960.1 | - | 2 | 887 | |||
AK173118.1 | - | 2 | 945 | |||
AY237008.1 | - | 1 | 505 | |||
BC010598.1 | - | 2 | 907 | 3498778 | ||
BC022907.1 | - | 2 | 894 |