Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153553.4 | NP_705781.1 | 7 | 1190 | ||
GenBank mRNA | AB049835.1 | - | 7 | 1085 | ||
AK032607.1 | - | 5 | 1703 | |||
AY730724.1 | - | 7 | 1217 | |||
BC129861.1 | - | 7 | 1126 | 40110003 |