Gene Symbol: Npas4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153553.4 | NP_705781.1 | 7 | 2047 | ||
GenBank mRNA | AB049835.1 | - | 7 | 1942 | ||
AK032607.1 | - | 5 | 2560 | |||
AY730724.1 | - | 7 | 2074 | |||
BC059815.1 | - | 5 | 1199 | |||
BC079544.1 | - | 5 | 1199 | |||
BC129861.1 | - | 7 | 1983 | 40110003 |