Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sbsn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172205.3 | NP_757342.2 | 1 | 1350 | ||
GenBank mRNA | AK081760.1 | - | 1 | 1287 | ||
AY115494.1 | - | 1 | 1343 | |||
BC029705.1 | - | 1 | 193 | |||
BC139385.1 | - | 1 | 1342 | 8862407 |