Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LGMN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008530.2 | NP_001008530.1 | 15 | 1996 | ||
NM_005606.6 | NP_005597.3 | 14 | 1878 | |||
XM_005267862.3 | XP_005267919.1 | 13 | 1693 | |||
XM_005267863.3 | XP_005267920.1 | 12 | 1625 | |||
XM_011536970.1 | XP_011535272.1 | 13 | 1796 | |||
XM_017021463.1 | XP_016876952.1 | 14 | 1688 | |||
XM_017021464.1 | XP_016876953.1 | 14 | 1793 | |||
XM_017021465.1 | XP_016876954.1 | 14 | 1682 | |||
XM_017021466.1 | XP_016876955.1 | 13 | 1600 | |||
XM_017021467.1 | XP_016876956.1 | 12 | 1543 | |||
GenBank mRNA | AK225822.1 | - | 14 | 1769 | ||
AK291534.1 | - | 14 | 1803 | |||
AK291560.1 | - | 14 | 1803 | |||
BC003061.2 | - | 14 | 1767 | 3504506 | ||
BC013678.1 | - | 5 | 835 | |||
BX161380.1 | - | 13 | 1613 | |||
BX161422.1 | - | 12 | 1542 | |||
BX161434.1 | - | 15 | 1906 | |||
D55696.1 | - | 13 | 1681 |