Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss11e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172880.2 | NP_766468.1 | 5 | 489 | ||
GenBank mRNA | AK037235.1 | - | 5 | 491 | ||
AK162701.1 | - | 5 | 486 | |||
AY787860.1 | - | 5 | 497 | |||
BC115432.1 | - | 5 | 483 | 40094164 | ||
BC115433.1 | - | 5 | 483 | 40094170 |