Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lpcat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173014.1 | NP_766602.1 | 4 | 662 | ||
GenBank mRNA | AB244716.1 | - | 4 | 557 | ||
AK039431.1 | - | 4 | 662 | |||
AK183634.1 | - | 2 | 56 | |||
AK192887.1 | - | 1 | 24 | |||
BC144805.1 | - | 4 | 617 |