Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFD1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035247.2 | NP_001030324.2 | 10, 11 | 907 | ||
NM_005659.6 | NP_005650.2 | 10, 11 | 907 | |||
XM_011530361.2 | XP_011528663.1 | 10, 11 | 1033 | |||
GenBank mRNA | AF141201.1 | - | 9, 10 | 777 | ||
AJ239058.1 | - | 9, 10 | 777 | |||
AK128120.1 | - | 5, 6 | 2798 | |||
AK225877.1 | - | 10, 11 | 864 | |||
AK304734.1 | - | 10, 11 | 877 | |||
AK315998.1 | - | 10, 11 | 858 | |||
AY101594.1 | - | 10, 11 | 834 | |||
BC001049.2 | - | 10, 11 | 854 | 3507963 | ||
BC005087.1 | - | 10, 11 | 855 | 3830774 | ||
BX648483.1 | - | 10, 11 | 839 | |||
CR456607.1 | - | 10, 11 | 791 | |||
U64444.1 | - | 10, 11 | 963 |