Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 9430077A04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK035035.1 | - | 1 | 1054 | ||
AK051584.1 | - | 994 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | NM_177134.1 | - | 1054 | Chr. - - [-] on Build |