Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lypd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177139.5 | NP_796113.1 | 3 | 401 | ||
GenBank mRNA | AK053620.1 | - | 4 | 532 | ||
BB619054.1 | - | 3 | 368 | |||
BB653059.1 | - | 3 | 362 | |||
BC070462.1 | - | 3 | 422 | 30650880 |