Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Islr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161535.1 | NP_001155007.1 | 2 | 3435 | ||
NM_001161536.1 | NP_001155008.1 | 4 | 3616 | |||
NM_001161537.1 | NP_001155009.1 | 3 | 3607 | |||
NM_001161538.1 | NP_001155010.1 | 4 | 3384 | |||
NM_001161539.1 | NP_001155011.1 | 4 | 3346 | |||
NM_001161540.1 | NP_001155012.1 | 3 | 3302 | |||
NM_001161541.1 | NP_001155013.1 | 3 | 3193 | |||
NM_177193.5 | NP_796167.2 | 3 | 3426 | |||
GenBank mRNA | AK043849.1 | - | 1 | 1260 | ||
AK047348.1 | - | 1 | 2914 | |||
AK129367.1 | - | 3 | 3113 | |||
BC059068.1 | - | 3 | 3279 | 6825665 | ||
BC096531.1 | - | 2 | 3435 | 6826287 |