Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ARPC1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005720.3 | NP_005711.1 | 10 | 1517 | ||
XM_006715825.1 | XP_006715888.1 | 10 | 1528 | |||
XM_006715826.1 | XP_006715889.1 | 10 | 1512 | |||
GenBank mRNA | AY007111.1 | - | 8 | 1347 | ||
BC002562.2 | - | 10 | 1450 | 3138319 | ||
BC002988.1 | - | 10 | 1488 | 2821335 |