Search Thermo Fisher Scientific
Gene Symbol: Ccdc158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177230.3 | NP_796204.1 | 6 | 675 | ||
GenBank mRNA | AK029997.1 | - | 6 | 675 | ||
AK076225.1 | - | 6 | 701 | |||
AK160895.1 | - | 6 | 699 | |||
BC137976.1 | - | 6 | 644 | 8860995 | ||
BC145072.1 | - | 6 | 645 |