Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177271.3 | NP_796245.2 | 2 | 976 | ||
GenBank mRNA | AK045645.1 | - | 2 | 528 | ||
AK087498.1 | - | 2 | 978 | |||
AK132444.1 | - | 2 | 950 | |||
BC145672.1 | - | 2 | 750 | 40130975 | ||
BC145674.1 | - | 2 | 750 | 40130977 |