Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024652.4 | NP_078928.3 | 34 | 6411 | ||
XM_005254979.3 | XP_005255036.1 | 20 | 4069 | |||
XM_011522012.2 | XP_011520314.1 | 36 | 6685 | |||
XM_011522013.2 | XP_011520315.1 | 34 | 6394 | |||
XM_011522014.2 | XP_011520316.1 | 33 | 6507 | |||
XM_011522015.2 | XP_011520317.1 | 29 | 7204 | |||
XM_011522016.2 | XP_011520318.1 | 32 | 5886 | |||
XM_011522017.2 | XP_011520319.1 | 20 | 4076 | |||
XM_011522018.2 | XP_011520320.1 | 20 | 4071 | |||
XM_017022570.1 | XP_016878059.1 | 24 | 4658 | |||
GenBank mRNA | AB058693.1 | - | 20 | 4116 | ||
AK026772.1 | - | 3 | 786 | |||
AK054662.1 | - | 8 | 1858 | |||
AK058028.1 | - | 14 | 2959 | |||
AK094556.1 | - | 4 | 1539 | |||
AK131075.1 | - | 15 | 3344 | |||
AK225575.1 | - | 3 | 786 | |||
BC005408.1 | - | 10 | 2230 | |||
BC020379.1 | - | 10 | 2230 | |||
BC066655.1 | - | 5 | 1324 | |||
BC110293.1 | - | 3 | 970 | |||
BC144705.1 | - | 32 | 5799 | |||
BC146769.1 | - | 20 | 4116 | |||
BC171802.1 | - | 33 | 6278 |