Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ifne | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177348.2 | NP_796322.1 | 1 | 483 | ||
GenBank mRNA | AY190044.1 | - | 1 | 483 | ||
BC104376.2 | - | 1 | 482 | 40050053 | ||
BC104377.2 | - | 1 | 482 | 40050054 |