Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Map3k9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177395.5 | NP_796369.2 | 8 | 2439 | ||
GenBank mRNA | AK053843.1 | - | 8 | 2280 |