Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mogat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177448.4 | NP_803231.1 | 6 | 1524 | ||
GenBank mRNA | AK049560.1 | - | 6 | 1488 | ||
AK133854.1 | - | 6 | 1530 | |||
AY157609.1 | - | 6 | 1481 | |||
BC052831.1 | - | 6 | 1528 | 30041797 |