Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195265.1 | NP_001182194.1 | 16 | 3005 | ||
GenBank mRNA | AK033653.1 | - | 6 | 900 | ||
BC096488.1 | - | 6 | 1121 | |||
BC147398.1 | - | 3 | 808 | 9056452 | ||
BC147399.1 | - | 3 | 808 | 9056443 | ||
BC172054.1 | - | 3 | 808 | |||
KF041446.1 | - | 17 | 2971 | |||
KF041447.1 | - | 17 | 3000 | |||
KF041448.1 | - | 14 | 2785 | |||
KF041449.1 | - | 15 | 2818 | |||
KF041450.1 | - | 16 | 2910 | |||
KF041451.1 | - | 17 | 2791 |