Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177622.3 | NP_808290.1 | 4 | 1116 | ||
GenBank mRNA | AK038563.1 | - | 4 | 1117 | ||
AK134150.1 | - | 4 | 1094 | |||
AK136304.1 | - | 4 | 1104 | |||
AK154058.1 | - | 4 | 1094 | |||
BC141018.1 | - | 4 | 1189 | 9055630 |