Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp874b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001076791.2 | NP_001070259.1 | 4 | 910 | ||
GenBank mRNA | AK079330.1 | - | 2 | 617 | ||
AK137581.1 | - | 4 | 888 |
Gene Symbol: Zfp874a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177712.4 | NP_808380.1 | 4 | 913 | ||
GenBank mRNA | AK049183.1 | - | 4 | 901 | ||
AK145787.1 | - | 4 | 913 | |||
BC058540.1 | - | 1 | 102 | |||
BC080824.1 | - | 992 | 30458333 |