Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318889.1 | NP_001305818.1 | 2 | 358 | ||
NM_005816.4 | NP_005807.1 | 2 | 358 | |||
NM_198196.2 | NP_937839.1 | 2 | 358 | |||
NR_134917.1 | 2 | 358 | ||||
XM_005247063.3 | XP_005247120.1 | 2 | 319 | |||
XM_006713469.3 | XP_006713532.1 | 2 | 319 | |||
XM_006713470.3 | XP_006713533.1 | 2 | 319 | |||
XM_017005521.1 | XP_016861010.1 | 2 | 319 | |||
XM_017005522.1 | XP_016861011.1 | 2 | 319 | |||
GenBank mRNA | AK290719.1 | - | 2 | 244 | ||
AK315237.1 | - | 2 | 240 | |||
AL833681.1 | - | 2 | 205 | |||
AW341041.1 | - | 2 | 338 | |||
BC020749.1 | - | 2 | 289 | 4669495 | ||
BM561433.1 | - | 2 | 358 | |||
DB130968.1 | - | 2 | 344 |