Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005819.5 | NP_005810.1 | 2 | 522 | ||
XM_011509058.1 | XP_011507360.1 | 2 | 522 | |||
XM_011509059.1 | XP_011507361.1 | 2 | 318 | |||
XM_017000007.1 | XP_016855496.1 | 2 | 318 | |||
GenBank mRNA | AJ002078.1 | - | 2 | 74 | ||
AK312440.1 | - | 2 | 309 | |||
BC009944.2 | - | 2 | 245 | 4122224 | ||
BU902578.1 | - | 2 | 312 | |||
CR541856.1 | - | 2 | 74 | |||
CR541873.1 | - | 2 | 74 |