Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005829.4 | NP_005820.1 | 2, 3 | 597 | ||
NR_023361.1 | 2, 3 | 597 | ||||
NR_037582.1 | 2, 3 | 593 | ||||
GenBank mRNA | AK222698.1 | - | 2, 3 | 189 | ||
AK294712.1 | - | 2, 3 | 189 | |||
AK312467.1 | - | 2, 3 | 221 | |||
AL110108.1 | - | 2, 3 | 185 | |||
BC002785.2 | - | 2, 3 | 182 | 3614033 | ||
BC007773.2 | - | 2, 3 | 181 | |||
BC010020.2 | - | 2, 3 | 209 | 2959670 | ||
DA777624.1 | - | 2, 3 | 236 | |||
DC340463.1 | - | 2, 3 | 206 | |||
X99459.1 | - | 2, 3 | 191 |
Gene Symbol: C15orf38-AP3S2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199058.1 | NP_001185987.1 | 6, 7 | 919 | ||
GenBank mRNA | AK000495.1 | - | 6, 7 | 811 |