Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scai | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178778.3 | NP_848893.2 | 18 | 2233 | ||
GenBank mRNA | AK044772.1 | - | 18 | 2229 | ||
AK046875.1 | - | 3 | 744 |