Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trim65 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178802.4 | NP_848917.2 | 6 | 1301 | ||
GenBank mRNA | AK028857.1 | - | 6 | 1190 | ||
AK047439.1 | - | 6 | 1301 | |||
AK050080.1 | - | 6 | 1213 | |||
AK079982.1 | - | 5 | 1383 | |||
BC118954.1 | - | 6 | 1056 | |||
BC118955.1 | - | 6 | 1056 |