Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005862.2 | NP_005853.2 | 3 | 390 | ||
XM_011512329.2 | XP_011510631.1 | 3 | 364 | |||
XM_011512331.2 | XP_011510633.1 | 3 | 399 | |||
XM_017005523.1 | XP_016861012.1 | 3 | 365 | |||
GenBank mRNA | AB210023.1 | - | 3 | 355 | ||
AK309934.1 | - | 3 | 365 | |||
AU132652.1 | - | 3 | 390 | |||
BC064699.1 | - | 3 | 365 | 30342305 | ||
CR749247.1 | - | 3 | 365 | |||
Z75330.1 | - | 3 | 498 |