Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SF3A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005877.5 | NP_005868.1 | 2 | 253 | ||
GenBank mRNA | AK303273.1 | - | 2 | 228 | ||
AU100212.1 | - | 2 | 230 | |||
BC001976.1 | - | 2 | 193 | 3459964 | ||
BC007684.2 | - | 2 | 220 | 3634035 | ||
BI827130.1 | - | 2 | 207 | |||
CR456575.1 | - | 2 | 196 | |||
DB445415.1 | - | 2 | 254 | |||
X85237.1 | - | 2 | 195 |