Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem145 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183311.2 | NP_899134.2 | 12 | 1019 | ||
GenBank mRNA | AK081054.1 | - | 12 | 1020 |