Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cfap36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025740.3 | NP_080016.1 | 6, 7 | 685 | ||
GenBank mRNA | AK010468.1 | - | 6, 7 | 712 | ||
AK075858.1 | - | 6, 7 | 686 | |||
BC005731.1 | - | 6, 7 | 655 | 3595965 |