Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ugp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290634.1 | NP_001277563.1 | 6 | 826 | ||
NM_139297.6 | NP_647458.1 | 6 | 1016 | |||
GenBank mRNA | AF424698.1 | - | 6 | 864 | ||
AK033445.1 | - | 6 | 863 | |||
AK153885.1 | - | 5 | 1959 | |||
AK218540.1 | - | 3 | 184 | |||
BC023810.1 | - | 6 | 867 | 5353993 | ||
BC025585.1 | - | 6 | 771 | 5324928 | ||
BC026626.1 | - | 5 | 553 | |||
BC061208.1 | - | 6 | 903 | 30250244 |