Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cecr5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144815.2 | NP_659064.1 | 2 | 152 | ||
GenBank mRNA | AK044228.1 | - | 2 | 352 | ||
AK140387.1 | - | 2 | 173 | |||
BC014705.1 | - | 2 | 161 | 4238201 |