Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BC017643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252548.1 | NP_001239477.1 | 7 | 1306 | ||
NM_001252549.1 | NP_001239478.1 | 7 | 1152 | |||
NM_001252550.1 | NP_001239479.1 | 6 | 1252 | |||
NM_001254735.1 | NP_001241664.1 | 7 | 1229 | |||
NM_144832.2 | NP_659081.1 | 7 | 1326 | |||
GenBank mRNA | AK143743.1 | - | 5 | 2343 | ||
AK145884.1 | - | 7 | 1290 | |||
AK158402.1 | - | 7 | 1266 | |||
AK159453.1 | - | 7 | 1284 | |||
AK168104.1 | - | 7 | 1284 | |||
AK170767.1 | - | 7 | 1274 | |||
AK213666.1 | - | 1 | 365 | |||
BC017643.1 | - | 7 | 1282 | 4503381 | ||
BC026616.1 | - | 7 | 1217 | 4977003 | ||
BC027406.1 | - | 7 | 1111 | 4950930 |