Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2e2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144839.1 | NP_659088.1 | 6 | 1160 | ||
GenBank mRNA | AK162602.1 | - | 55 | |||
BC016265.1 | - | 6 | 1160 | 4923869 |