Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6330416G13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144905.3 | NP_659154.2 | 10 | 1682 | ||
GenBank mRNA | AK030466.1 | - | 10 | 1812 | ||
AK031016.1 | - | 10 | 1686 | |||
AK078066.1 | - | 1 | 352 | |||
AK138872.1 | - | 8 | 1499 | |||
AK144990.1 | - | 11 | 1859 | |||
BC022599.1 | - | 8 | 1536 | 4159235 | ||
BC023748.1 | - | 10 | 1854 | 5353894 |