Gene Symbol: Wbscr17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145218.3 | NP_660253.2 | 8 | 2203 | ||
GenBank mRNA | AB175684.1 | - | 8 | 1281 | ||
AF467979.1 | - | 8 | 1484 | |||
AK035817.1 | - | 8 | 1957 | |||
AK048758.1 | - | 8 | 1948 | |||
AK051281.1 | - | 8 | 2603 | |||
BC052469.1 | - | 8 | 1599 | 6810287 | ||
BC069177.1 | - | 8 | 2203 |