Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Nipal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145469.5 | NP_663444.2 | 3 | 267 | ||
GenBank mRNA | AK033975.1 | - | 3 | 268 | ||
AK038556.1 | - | 3 | 408 | |||
AK079085.1 | - | 3 | 700 | |||
AK132530.1 | - | 3 | 295 | |||
BC016107.1 | - | 3 | 290 | 4921508 | ||
BC030399.1 | - | 3 | 784 | 4166825 | ||
BC058207.1 | - | 3 | 433 | 5361417 |