Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NET1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047160.2 | NP_001040625.1 | 5, 6 | 678 | ||
NM_005863.4 | NP_005854.2 | 3, 4 | 589 | |||
GenBank mRNA | AB209852.1 | - | 5, 6 | 624 | ||
AJ010046.1 | - | 3, 4 | 522 | |||
AK024919.1 | - | 3, 4 | 529 | |||
AK299335.1 | - | 5, 6 | 634 | |||
AK315566.1 | - | 3, 4 | 531 | |||
BC010285.1 | - | 3, 4 | 520 | 3048980 | ||
BC053553.1 | - | 5, 6 | 630 | 6137683 | ||
U02081.1 | - | 1, 2 | 110 |