Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145482.3 | NP_663457.2 | 11 | 1534 | ||
GenBank mRNA | AK135996.1 | - | 2 | 600 | ||
AK137485.1 | - | 2 | 2327 | |||
AY037804.1 | - | 11 | 1534 | |||
BC029101.1 | - | 11 | 2049 | |||
BY701902.1 | - | 2 | 73 |