Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cstf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037326.2 | NP_001032403.1 | 2 | 466 | ||
NM_145529.3 | NP_663504.1 | 3 | 348 | |||
NM_177253.4 | NP_796227.1 | 3 | 348 | |||
GenBank mRNA | AK028899.1 | - | 3 | 293 | ||
AK083093.1 | - | 2 | 332 | |||
AK159180.1 | - | 3 | 151 | |||
AK164375.1 | - | 1 | 4532 | |||
BC003241.1 | - | 3 | 309 | 3498424 | ||
BC058605.1 | - | 3 | 300 |