Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmprss11d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145561.2 | NP_663536.1 | 3 | 256 | ||
GenBank mRNA | AB053953.1 | - | 3 | 290 | ||
AF448809.1 | - | 3 | 235 | |||
BC020151.1 | - | 3 | 256 | 4021767 |