Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAMPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005746.2 | NP_005737.1 | 2 | 469 | ||
XM_005250100.2 | XP_005250157.1 | 3 | 450 | |||
GenBank mRNA | AK292851.1 | - | 2 | 229 | ||
BC072439.1 | - | 2 | 242 | 6023159 | ||
BC106046.1 | - | 2 | 265 | 6161081 | ||
U02020.1 | - | 2 | 188 |