Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vstm2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290539.1 | NP_001277468.1 | 5 | 2541 | ||
NM_145967.2 | NP_666079.2 | 5 | 2663 | |||
GenBank mRNA | AK134954.1 | - | 2 | 1916 | ||
AK137363.1 | - | 5 | 2583 | |||
BC027127.1 | - | 5 | 2614 | 5361611 |