Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282055.1 | NP_001268984.1 | 11, 12 | 1638 | ||
GenBank mRNA | AK088950.1 | - | 11, 12 | 1610 | ||
AK129220.1 | - | 11, 12 | 1611 | |||
BC026492.1 | - | 4, 5 | 583 | 4505095 | ||
BC092240.1 | - | 10, 11 | 1549 | 30662573 |