Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc45a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177628.2 | NP_001171099.1 | 3 | 1211 | ||
NM_145977.2 | NP_666089.1 | 3 | 1292 | |||
GenBank mRNA | AK035428.1 | - | 3 | 1161 | ||
AK135655.1 | - | 3 | 483 | |||
BC031381.1 | - | 3 | 1292 | 5050610 | ||
BC034084.1 | - | 1 | 541 |